Order Entry
Austria
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
Irritant
Corrosive
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Proteomics Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Cloud Point:
    Report
  • Colour (APHA):
    ≤100
  • Peroxides:
    None detectable
  • pH (5 %, Water) @25 °C:
    6,0 - 8,0
  • Protease:
    None detectable

Specification Test Results

Related Information

Frequently Bought Together