Order Entry
Austria
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 98% for molecular biology DNase-, RNase-, protease-free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 98% for molecular biology DNase-, RNase-, protease-free
Certificates
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 98% for molecular biology DNase-, RNase-, protease-free
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear viscous liquid
  • Water:
    ≤0.2 %
  • HPLC:
    ≥97.5 %
  • pH:
    6 to 8 (5 % in water)
  • Dnase,Rnase,Protease activity:
    Not detected
  • Heavy metals (as Pb):
    ≤5 ppm
  • Peroxides:
    ≤0.01 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together