ContactUsLinkComponent
Polyethylene glycol 400, GPR RECTAPUR®
Polyethylene glycol 400, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Polyethylene glycol 400, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Identification:
    Passes test
  • Acid value (mg KOH/g):
    Max. 0.2
  • Density (20/4):
    1.120 - 1.130
  • Hydroxyl value:
    200 - 300