Polyethylene glycol 3,000 for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
:3000 g/mol
Melting Pt:55…58 °C
Density:1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-038-2
Specification Test Results
- Solubility:550 g/l (20 °C)
- Melting Point:55 - 58 °C
- Molar Mass:3000 g/mol
- Bulk Density:400 - 500 kg/m3
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:270 °C
- Density:1.21 g/cm3 (20 °C)
- pH:4 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:420 °C
- Viscosity kinematic:69 - 92 mm2/s (20 °C)
Specification Test Results
Frequently Bought Together