Order Entry
Austria
ContactUsLinkComponent
Polyethylene glycol 3,000, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Catalog # 8.17019.5000
CAS Number:  
Polyethylene glycol 3,000, EMPROVE® ESSENTIAL Ph. Eur., SAFC®
Catalog # 8.17019.5000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :3000 g/mol
Melting Pt:55…58 °C
Density:1.21 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH