ContactUsLinkComponent
Poly(dimethylsiloxane-co-methylphenylsiloxane), 50 cSt (25°C), Sigma-Aldrich®
  SIAL378445-50ML
 :  Merck
 :  
Poly(dimethylsiloxane-co-methylphenylsiloxane), 50 cSt (25°C), Sigma-Aldrich®
  SIAL378445-50ML
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370
 :
 :∼35 °C (1013 hPa)
 :0.982 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
 :>101.1 °C (closed cup)
 :Ambient
 :63148-52-7