J.T.Baker® BAKER INSTRA-ANALYZED®, Silver ICP Standard Solution, 10000 mg/L Ag in dil. Nitric Acid
Catalog # 5745.0100
Supplier: Avantor
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Element:Silver
- Symbol:Ag