Order Entry
Austria
ContactUsLinkComponent
J.T.Baker®, Silver Standard Solution for AAS, 1000 mg/L Ag in 0,5 M Nitric Acid
J.T.Baker®, Silver Standard Solution for AAS, 1000 mg/L Ag in 0,5 M Nitric Acid
Catalog # 6821.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Silver Standard Solution for AAS, 1000 mg/L Ag in 0,5 M Nitric Acid
Catalog # 6821.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Silver
  • Symbol:
    Ag

Specifications

About this item