Order Entry
Austria
ContactUsLinkComponent
 
Lead standard solution, 10000 mg/L Pb in dil. nitric acid (from Pb(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70372.0100
Supplier:  Merck
CAS Number:  
Lead standard solution, 10000 mg/L Pb in dil. nitric acid (from Pb(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70372.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item

Certifications: Traceable to SRM from NIST.