Order Entry
Austria
ContactUsLinkComponent
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Catalog # 1.19812.0500
Supplier:  Merck
CAS Number:  
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Catalog # 1.19812.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Ammonium standard solution
  • M-Clarity quality level:
    MQ100
  • Contents:
    NH₄Cl in H₂O

Specifications

About this item

Certifications: Traceable to SRM from NIST.