Order Entry
Austria
ContactUsLinkComponent
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al(NO₃)₃.9H₂O) TraceCERT® standard for ICP, Supelco®
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al(NO₃)₃.9H₂O) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL61935-100ML-F
Supplier:  Merck
CAS Number:  
Aluminium standard solution, 1000 mg/L Al in dil. nitric acid (from Al(NO₃)₃.9H₂O) TraceCERT® standard for ICP, Supelco®
Catalog # SUPL61935-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the certificate.