Orders LinkContactUsLinkComponent
Precision heating bath
Precision heating bath
Supplier:   GESTIGKEIT HARRY
Precision heating bath
Supplier:  GESTIGKEIT HARRY
Certificates
Precision heating bath
Precision heating bath
2 Options Available Below
Product Details & Documents
Precision heating baths which can be used for fluid, oil, air or as a sandbath for continuous operation. They are very well suited for use with rotary evaporators, heating is provided by a tubular radiator.
  • Made of stainless steel with spray-painted base
The appliances are controlled by means of a microprocessor PID-controller in connection with a low-mass sensor. The actual-value temperature is always on view. The preset temperature can be seen by pushing a button and can also be adjusted with the up/down key.
Product Family Options
Product Information
  • Type
    Temp.
    Capacity
    Power
    W×D×H
    Weight
    Availability
    Price
  • GESTPZ26-4
    Digital heating bath for fluid, air or sandbath, 230 V
    Digital heating bath for fluid, air or sandbath, 230 V
    PZ 26-4 heating bath
    400 °C
    5 L
    1200 W
    310×310×280 mm
    6,8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 460,00 €
    1 460,00 €
    1 * 1 ST
       1 460,00 €
  • GESTPZ26-7
    Digital heating bath for dry, air or sandbath, 230 V
    Digital heating bath for dry, air or sandbath, 230 V
    PZ 26-7 heating bath
    700 °C
    5 L
    1200 W
    310×310×280 mm
    6,8 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €