Order Entry
Austria
ContactUsLinkComponent
VWR®, Bovine serum albumin (BSA)
VWR®, Bovine serum albumin (BSA)
  422381B
 :  
undefined
VWR®, Bovine serum albumin (BSA)
  422381B
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Endotoxin level:
    Low
  • Colour:
    light yellow

 

 

Produced by a heat shock process using sodium caprylate as a stabiliser with additional treatments including dialysis and deionisation.

  • Low endotoxin