Order Entry
Austria
ContactUsLinkComponent
VWR®, Chicken native albumin (from egg)
VWR®, Chicken native albumin (from egg)
Catalog # 20771.236
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Chicken native albumin (from egg)
Catalog # 20771.236
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • CAS No.:
    9006-59-1
  • Protein/peptide type:
    Native
  • Source:
    Egg
  • Species:
    Chicken
  • Storage conditions:
    Refrigerator
  • Form:
    Powder
  • Protein/peptide name:
    Albumin
  • Grade:
    Technical Grade

Specifications

About this item