VWR®, Gelatin
Supplier: VWR Chemicals
Certificates
About this item
- Pale yellow powder
Specifications
- CAS No.:9000-70-8
- Storage conditions:Ambient
- Form:Powder
- Protein synonyms:Gelatine
- Protein/peptide name:Gelatin
- Grade:Ph. Eur.
Specifications
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
24360_GELATINE POWDER PH.EUR_BSE_Rev1
BSE/TSE Certificates
spec_VWRC24360.233 spec_VWRC24360.233 spec_VWRC24360.368 spec_VWRC24360.368
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together