Order Entry
Austria
ContactUsLinkComponent
Xanthine oxidase (from buttermilk)
Xanthine oxidase (from buttermilk)
Supplier:  Merck
Certificates
Xanthine oxidase (from buttermilk)
Supplier:  Merck

Specifications

  • CAS No.:
    9002-17-9
  • Source:
    Buttermilk
  • Enzyme name:
    Xanthine oxidase

Specifications

Frequently Bought Together