Order Entry
Austria
ContactUsLinkComponent
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, 4x crystallised
Toxic
Health
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, 4x crystallised
Catalog # A1577.1000
Supplier:  PanReac AppliChem
CAS Number:  
Toxic
Health
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution, 4x crystallised
Catalog # A1577.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :79-06-1
MDL Number:MFCD00080848