Order Entry
Austria
ContactUsLinkComponent
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution
Catalog # A4983.0500
Supplier:  PanReac AppliChem
CAS Number:  
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution
Catalog # A4983.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :79-06-1
MDL Number:MFCD00080848