Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acrylamide-Bis (32:1)
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution, 4x crystallised
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution, 4x crystallised
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution, 4x crystallised
A0951.1000
:
PanReac AppliChem
:
Acrylamide : Bisacrylamide (29:1) 30% in aqueous solution, 4x crystallised
A0951.1000
:
PanReac AppliChem
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
3426
:
6.1,III
:
GHS06
:
GHS08
:
79-06-1
:
MFCD00080848