DL-Sodium lactate 60% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:72-17-3
:312-85-6
Formula:H₃CCH(OH)COONa
MDL Number:MFCD00065400
:112.06 g/mol
Density:1.27…1.32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-772-0
Frequently Bought Together