Order Entry
Austria
ContactUsLinkComponent
Lactic acid 90% ≥90% in aqueous solution, GPR RECTAPUR®, pure
Corrosive
Lactic acid 90% ≥90% in aqueous solution, GPR RECTAPUR®, pure
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Lactic acid 90% ≥90% in aqueous solution, GPR RECTAPUR®, pure
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 90 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.1 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together