Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories for safety storage cabinets for toxic substances
Accessories for safety storage cabinets for toxic substances
Accessories for safety storage cabinets for toxic substances
DUEH20-81045-124
:
DUPERTHAL
Accessories for safety storage cabinets for toxic substances
DUEH20-81045-124
:
DUPERTHAL
Description:
Additional lower shelf, perforated and 3 mm galvanised, W×D×H: 935×365×63 mm, 4 kg
Pk:
1
For:
Toxic substances cabinet L