Order Entry
Austria
ContactUsLinkComponent
 
Automated stainer, Cellstain® 15
Catalog # THARJC665-30
Supplier:  THARMAC GmbH
undefined
Automated stainer, Cellstain® 15
Catalog # THARJC665-30
Supplier:  THARMAC GmbH

Specifications

  • No. of slides:
    30
  • Description:
    Basic stainer with 15 troughs (500 ml) and slide holder
  • Seq:
    02
  • Environmentally Preferable:
  • W×D×H:
    950×280×350 mm
  • Available stations:
    15
  • Weight:
    16 kg
  • Pk:
    1

Specifications

About this item

The Cellstain 15 is a fully automated stainer for Papanicolaou, HE and other stainings.

  • 20 programmable procedures in the memory
  • Low transfer of reagents through the slide holder
  • Small format fitting under any existing fume hood
  • Agitation and drain time are freely selectable
  • Moveable slide holder to enable the user to program a large number of staining programs
Ordering information: Supplied with 15 staining troughs, slide holder and shortened trough for loading and unloading.