Agarose, Sigma-Aldrich®
Catalog # ROCH11685678001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
:
Melting Pt:60…90 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:232-731-8
CAS Number:9012-36-6
MDL Number:MFCD00081294