Agarose IEF
Catalog # 17-0468-01
Supplier: Cytiva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
About this item
:
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
CAS Number:9012-36-6
MDL Number:MFCD00081294