Agarose, Electran, high resolution, low melting for electrophoresis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Moisture content:Max. 10 %
- Sulfate content:Max. 0.15 %
- Gel strength (at 4%):Min. 500 g/cm²
- EEO, -Mr:Max. 0.15
- Gelling temperature (at 4%):Max. 35 °C
- Melting temperature (at 4%):Max. 65 °C
- Appearance:Passes
- Ash content:Max. 1 %
- DNA binding:Passes
- RNase / DNase:Not detected
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC436552V spec_VWRC436552V spec_VWRC436553W spec_VWRC436553W
Product specification
VWR® Products for Electrophoresis
Product Literature
Frequently Bought Together