Order Entry
Austria
Orders LinkContactUsLinkComponent
Agarose

Agarose

Supplier:   PanReac AppliChem
Agarose
Supplier:  PanReac AppliChem
Certificates
Agarose

3 Options Available Below

Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 220,00 €
    220,00 €
    1 * 100 g
       220,00 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 518,00 €
    518,00 €
    1 * 250 g
       518,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 884,00 €
    884,00 €
    1 * 500 g
       884,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.