Order Entry
Austria
Orders LinkContactUsLinkComponent
Agar

Agar

5 kg
Catalog # OXOILP0011T
Agar
Catalog # OXOILP0011T
CAS Number:  9002-18-0
Agar
CAS Number:  9002-18-0
Price
2 920,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.