Order Entry
Austria
Orders LinkContactUsLinkComponent
Agar

Agar

Anbieter:   PanReac AppliChem
Agar
Anbieter:  PanReac AppliChem
Certificates
Agar

3 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Verfügbarkeit
    Preis
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 159,00 €
    159,00 €
    1 * 500 g
       159,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 295,00 €
    295,00 €
    1 * 1 kg
       295,00 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1 170,00 €
    1 170,00 €
    1 * 5 kg
       1 170,00 €