Order Entry
Austria
ContactUsLinkComponent
 
Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free
Catalog # M228-10ML-5PK
Supplier:  VWR Chemicals
CAS Number:  
undefined
Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free
Catalog # M228-10ML-5PK
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 KIT
  • Seq:
    2882
  • Pk Info:
    5× 10 ml

Specifications

Specification Test Results

  • DNase:
    NONE
  • Heavy Metals:
    0.001 %
  • Identification:
    PASS
  • Milliflex (SDA):
    NONE
  • Milliflex (TSA):
    NONE
  • Peroxide:
    1.0 umol/ml
  • Protease:
    NONE
  • Residue on Ignition:
    0.25 %
  • RNase:
    NONE

Specification Test Results

About this item

 :
 :1227.53 g/mol
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986