Order Entry
Austria
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Proteomics Grade peroxide-free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Proteomics Grade peroxide-free
Catalog # M236-10ML-5PK
Supplier:  VWR Chemicals
CAS Number:  
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Proteomics Grade peroxide-free
Catalog # M236-10ML-5PK
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    2882
  • Environmentally Preferable:
  • Pk Info:
    5× 10 ml ampoules

Specifications

Specification Test Results

  • Colour (APHA):
    ≤100
  • Peroxides-free:
    Pass/fail
  • pH (50%, Water, 25° C):
    6,0 - 8,0
  • Protease- free:
    Pass/fail

Specification Test Results

About this item

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
 :GHS05
Storage Temperature:Ambient
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505
Melting Pt:6
Flash Pt:251
Merck Index:13,06793