Order Entry
Austria
ContactUsLinkComponent
D-(-)-Fructose
D-(-)-Fructose
Catalog # MOLE34878019-100G
Supplier:  Molekula
CAS Number:  
undefined
D-(-)-Fructose
Catalog # MOLE34878019-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
 :
 :180.16 g/mol
Merck Index:13,04295
Boiling Pt:552 °C (1013 hPa)
Melting Pt:100…110 °C
Density:1.59 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-333-3
CAS Number:57-48-7
Formula:C₆H₁₂O₆
MDL Number:MFCD00148910