Order Entry
Austria
ContactUsLinkComponent
 
D-(-)-Fructose, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, FCC, extra pure suitable for use as excipient, SAFC®
 
undefined
D-(-)-Fructose, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, FCC, extra pure suitable for use as excipient, SAFC®

 

  • Environmentally Preferable:

 

 

  • Assay (α 20/D, 100 g/l, water, calc. on anhydrous substance):
    98.5 - 101.2 %
  • Assay (α 25/D, 100 g/l,water, calc. on anhydrous substance):
    98.0 - 102.0 %
  • Identity:
    passes test
  • Appearance of solution (EP):
    passes test
  • Appearance of solution (USP):
    passes test
  • Acidity or alkalinity:
    passes test
  • Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance):
    -93.5 - -91.0 °
  • Spec. rotation (α 25/D; 10 %; water; calc. on anhydrous substance):
    -90.8 - -87.2 °
  • Chloride (Cl):
    ≤ 0.01 %
  • Sulfate (SO₄):
    ≤ 0.02 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • As (Arsenic):
    ≤ 0.0001 %
  • Ba (Barium):
    passes test
  • Calcium and Magnesium (as Ca):
    ≤ 0.005 %
  • Cu (Copper):
    ≤ 0.0025 %
  • Pb (Lead):
    ≤ 0.00001 %
  • Zn (Zinc):
    ≤ 0.0025 %
  • Glucose (HPLC):
    ≤ 0.5 %
  • Foreign sugars:
    passes test
  • 5-Hydroxymethylfurfural and related substances:
    passes test
  • Residual solvents (ICH Q3C):
    excluded by the production process
  • Sulfated ash:
    ≤ 0.1 %
  • Water:
    ≤ 0.5 %

 

Frequently Bought Together