D-(-)-Fructose ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 103674Y
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (calculated on anhydrous):Min. 99.0 %
- IR Spectrum:Passes test
- Spec.opt.rot.(10 %;water)(on anhydrous):-93.5 to -91.0 °
- Glucose:Max. 0.5 %
- Ignition residue (SO4):Max. 0.05 %
- Insolubility in water:Max. 50 ppm
- Water:Max. 0.5 %
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- As (Arsenic):Max. 1 ppm
- Cd (Cadmium):Max. 1 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
- Pb (Lead):Max. 1 ppm
- Zn (Zinc):Max. 5 ppm
Specification Test Results
About this item
Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
:
:180.16 g/mol
Boiling Pt:552
Melting Pt:100…110 °C
Density:1.59 g/cm³ (20 °C. OECD 109)
Storage Temperature:Ambient
EINECS:200-333-3
CAS Number:57-48-7
Formula:C₆H₁₂O₆
MDL Number:MFCD00148910
Merck Index:13,04295