ContactUsLinkComponent
UV-Vis wavelenth standard
UV-Vis wavelenth standard
  MICSPQA-HOX-01
 :  MICROSOLV
 :  
undefined
UV-Vis wavelenth standard
  MICSPQA-HOX-01
 :  MICROSOLV
 :  
  • Pk:
    30 ml
  • Description:
    Wavelength calibration standard
Wavelength calibration standard solution for UV-Vis detectors is composed of 4% Holmium oxide in 10% Perchloric acid.