D-(+)-Trehalose dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
CAS Number:6138-23-4
:
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594
Molecular Weight:378.33 g/mol
Melting Pt:97…99 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:202-739-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together