Order Entry
Austria
ContactUsLinkComponent
D-(+)-Maltose monohydrate ≥92% (by anhydrous basis), GPR RECTAPUR®
D-(+)-Maltose monohydrate ≥92% (by anhydrous basis), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
D-(+)-Maltose monohydrate ≥92% (by anhydrous basis), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous):
    min. 92%
  • pH (20°C; 5 %):
    4.5 - 6.0
  • Spec.opt.rot.(10 %;water)(on anhydrous):
    135 - 139 °
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    4 - 7 %

Specification Test Results

Frequently Bought Together