Order Entry
Austria
ContactUsLinkComponent
D-(+)-Maltose monohydrate
D-(+)-Maltose monohydrate
Catalog # EHERC14734700
Supplier:  EHRENSTORFER
CAS Number:  
undefined
D-(+)-Maltose monohydrate
Catalog # EHERC14734700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :360.32 g/mol
Merck Index:13,05736
Melting Pt:160…165 °C
Storage Temperature:Ambient
EINECS:200-716-5
CAS Number:6363-53-7
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00149343