Order Entry
Austria
ContactUsLinkComponent
Dimethyl sulphoxide-[D₆] 100.0 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D₆] 100.0 atom % D for NMR spectroscopy
Catalog # ACRO320760075
CAS Number:  
undefined
Dimethyl sulphoxide-[D₆] 100.0 atom % D for NMR spectroscopy
Catalog # ACRO320760075
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Pk Info:
    10 x 0.75 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.95 %

Specification Test Results

About this item

 :
 :84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³
Flash Pt:88 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090