ContactUsLinkComponent
Mercury short arc reflector lamp
Mercury short arc reflector lamp
Catalog # ZEIS0000001313162
Supplier:  ZEISS
Mercury short arc reflector lamp
Catalog # ZEIS0000001313162
Supplier:  ZEISS
Specifications
  • For:
    X-Cite 120 illuminator
  • Description:
    Mercury short arc reflactor lamp module
  • Pk:
    1
Specifications
About this item
This mercury short arc reflector lamp is specially designed for X-Cite 120 illuminator system. The average lifetime performance of this lamp module is 2000 hours.