Chemical process indicators
About this item
Chemical process indicators provide a fast and consistent visual check on items that were sterilised by EO.
- Consistent and irreversible colour transition
- Permanent pressure-sensitive adhesive
- Indicator inks do not contain lead or other toxic heavy metals
Chemical indicator: Initial colour (purple) and signal colour (green)
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Certifications: Compliance to ISO 11140-1:2014.
3 Options Available Below
Product Details & Documents
Chemical process indicators provide a fast and consistent visual check on items that were sterilised by EO.
- Consistent and irreversible colour transition
- Permanent pressure-sensitive adhesive
- Indicator inks do not contain lead or other toxic heavy metals
Chemical indicator: Initial colour (purple) and signal colour (green)
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Certifications: Compliance to ISO 11140-1:2014.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- LengthWidthØAvailabilityPrice
- NAMSCPI-E01Plain dots, thickness 0,10 mm, 5000 indicators per rollPlain dots, thickness 0,10 mm, 5000 indicators per roll12,7 mmLog in for availability1 * 1 Rolle320,00 €
- NAMSCPI-E02Indicator labels for hand-held labeling systems, thickness 0,10 mm, 1500 indicators per rollIndicator labels for hand-held labeling systems, thickness 0,10 mm, 1500 indicators per roll25 mm12 mmLog in for availability1 * 1 Rolle109,00 €
- NAMSSP-E16Imprinted indicator labels for hand-held labeling systems, thickness 0,10 mm, 1500 indicators per rollImprinted indicator labels for hand-held labeling systems, thickness 0,10 mm, 1500 indicators per roll25 mm12 mmLog in for availability1 * 1.500 Rolle144,00 €