Order Entry
Austria
ContactUsLinkComponent
Methanol-[D₄], Sigma-Aldrich®
Flammable
Toxic
Health
Methanol-[D₄], Sigma-Aldrich®
Catalog # SIAL535435-10G
Supplier:  Merck
CAS Number:  
Flammable
Toxic
Health
Methanol-[D₄], Sigma-Aldrich®
Catalog # SIAL535435-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS06
 :GHS08
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637