Orders LinkContactUsLinkComponent
Magnetic stirrer, KMO 3 basic, IKA
Magnetic stirrer, KMO 3 basic, IKA
KMO 3 basic IKAMAG® magnetic stirrer, 50/60 Hz, EU/UK/CH-plug, 100/240 V
Catalog # 531-1342
Supplier:   IKA
Magnetic stirrer, KMO 3 basic, IKA
Catalog # 531-1342
Supplier:  IKA
Magnetic stirrer, KMO 3 basic, IKA
Magnetic stirrer, KMO 3 basic, IKA
KMO 3 basic IKAMAG® magnetic stirrer, 50/60 Hz, EU/UK/CH-plug, 100/240 V
Price
793,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compact magnetic stirrer with high quality, fast-response display and hardened glass work surface with a brilliant, clear white central area to easily view colour changes.
  • Torque display
  • Right and left rotation
  • Illuminated, flat stirring plate
  • Key lock button
  • Integrated USB interface (control via labworldsoft and use of Firmware Update Tool)
  • Recessed thread to accommodate support rod
Certifications: IP protection class according to DIN EN 60529: IP 42
Specifications
  • Description:
    KMO 3 basic IKAMAG® magnetic stirrer, 50/60 Hz, EU/UK/CH-plug
  • Voltage:
    100/240 V
  • Plate material:
    Glass
  • Cat. No.:
    531-1342
  • Speed display:
    LED
  • Model:
    KMO 3 basic IKAMAG®
  • Speed range:
    10 - 1500 min⁻¹
  • W×D×H:
    135 × 227 × 62,5 mm
  • Plate dimensions:
    140×120 mm
  • Max. stirring capacity H₂O:
    5 L
  • Weight:
    1,2 kg
Accessories
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4496400
    Cross-shaped
    20 mm
    20 mm
    Log in for availability
    1 * 5 ST - 28,00 €
Product Information
  • Catalog #
    Type
    Availability
    Price
  • 441-2121
    Set with IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 and TRIKA® 25 and 40 stirring bars
    Log in for availability
    1 * 1 SET - 146,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Holding rod, for fastening ETS-D5 or VT-5 to the support rod
    Holding rod, for fastening ETS-D5 or VT-5 to the support rod
    431-2888
    Log in for availability
    1 * 1 ST - 39,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4489100
    Cylindrical without pivot ring
    40 mm
    8 mm
    Log in for availability
    1 * 5 ST - 19,00 €
Product Information
  • Catalog #
    Length
    Ø
    Availability
    Price
  • 442-0902
    350 mm
    8 mm
    Log in for availability
    1 * 1 ST - 95,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Labworldsoft® 6 Starter
    Labworldsoft® 6 Starter
    IKAA20019397
    Log in for availability
    1 * 1 ST - 6 500,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4494000
    Cylindrical with removable pivot ring
    30 mm
    6 mm
    Log in for availability
    1 * 5 ST - 15,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4495400
    Elliptical
    40 mm
    20 mm
    Log in for availability
    1 * 5 ST - 56,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4488900
    Cylindrical without pivot ring
    25 mm
    8 mm
    Log in for availability
    1 * 5 ST - 17,80 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4495200
    Elliptical
    35 mm
    15 mm
    Log in for availability
    1 * 5 ST - 44,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4493200
    Cylindrical, made of rare earth magnet samarium-cobalt
    30 mm
    6 mm
    Log in for availability
    1 * 5 ST - 52,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Extension for support rod, H 16,3, for bath attachment over Ø 180 mm
    Extension for support rod, H 16,3, for bath attachment over Ø 180 mm
    IKAA25000622
    Log in for availability
    1 * 1 ST - 24,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4492800
    Cylindrical without pivot ring
    50 mm
    8 mm
    Log in for availability
    1 * 5 ST - 38,00 €
Product Information
  • Catalog #
    Availability
    Price
  • R 350 Universal clamp for clamping flask necks, condensers, etc. up to 11 cm diameter
    R 350 Universal clamp for clamping flask necks, condensers, etc. up to 11 cm diameter
    442-1133
    Log in for availability
    1 * 1 ST - 141,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4489000
    Cylindrical without pivot ring
    30 mm
    8 mm
    Log in for availability
    1 * 5 ST - 17,80 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4488700
    Cylindrical without pivot ring
    15 mm
    6 mm
    Log in for availability
    1 * 5 ST - 12,40 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4493800
    Cylindrical with removable pivot ring
    25 mm
    6 mm
    Log in for availability
    1 * 5 ST - 15,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4493000
    Cylindrical, made of rare earth magnet samarium-cobalt
    15 mm
    6 mm
    Log in for availability
    1 * 5 ST - 36,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4488600
    Cylindrical without pivot ring
    10 mm
    6 mm
    Log in for availability
    1 * 5 ST - 11,10 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4495600
    Elliptical
    50 mm
    20 mm
    Log in for availability
    1 * 5 ST - 78,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4493400
    Cylindrical, made of rare earth magnet samarium-cobalt
    50 mm
    8 mm
    Log in for availability
    1 * 5 ST - 63,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4496600
    Cross-shaped
    25 mm
    25 mm
    Log in for availability
    1 * 5 ST - 31,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4489200
    Cylindrical without pivot ring
    50 mm
    8 mm
    Log in for availability
    1 * 5 ST - 21,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4494400
    Cylindrical with removable pivot ring
    50 mm
    8 mm
    Log in for availability
    1 * 5 ST - 32,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Bosshead clamp
    Bosshead clamp
    442-2043
    Log in for availability
    1 * 1 ST - 42,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4496200
    Cross-shaped
    10 mm
    10 mm
    Log in for availability
    1 * 5 ST - 19,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4495000
    Elliptical
    30 mm
    15 mm
    Log in for availability
    1 * 5 ST - 36,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4494600
    Elliptical
    20 mm
    10 mm
    Log in for availability
    1 * 5 ST - 24,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread
    Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread
    442-2041
    Log in for availability
    1 * 1 ST - 88,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4498400
    Round with crosshead
    6 mm
    9 mm
    Log in for availability
    1 * 5 ST - 16,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4494800
    Elliptical
    25 mm
    12 mm
    Log in for availability
    1 * 5 ST - 28,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    IKAA20017366
    Log in for availability
    1 * 1 ST - 14 820,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4488800
    Cylindrical without pivot ring
    20 mm
    8 mm
    Log in for availability
    1 * 5 ST - 15,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4492200
    Cylindrical without pivot ring
    25 mm
    8 mm
    Log in for availability
    1 * 5 ST - 36,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4494200
    Cylindrical with removable pivot ring
    40 mm
    8 mm
    Log in for availability
    1 * 5 ST - 29,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4492600
    Cylindrical without pivot ring
    40 mm
    8 mm
    Log in for availability
    1 * 5 ST - 36,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4497000
    Cross-shaped
    38 mm
    38 mm
    Log in for availability
    1 * 5 ST - 48,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakers
    Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakers
    IKAA20003417
    Log in for availability
    1 * 1 ST - 22,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4496800
    Cross-shaped
    30 mm
    30 mm
    Log in for availability
    1 * 5 ST - 39,00 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4497200
    Double-ended
    37 mm
    20 mm
    Log in for availability
    1 * 5 ST - 69,50 €
Product Information
  • Catalog #
    Type
    Length
    Ø
    Availability
    Price
  • IKAA4497400
    Double-ended
    54 mm
    20 mm
    Log in for availability
    1 * 5 ST - 84,00 €