Order Entry
Austria
ContactUsLinkComponent
Soda lime, granular ACS +100 mesh, Fluka™
Corrosive
Soda lime, granular ACS +100 mesh, Fluka™
Catalog # 266434-500G
CAS Number:  
Corrosive
Soda lime, granular ACS +100 mesh, Fluka™
Catalog # 266434-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Form:
    granular
  • Loss on drying (200°C):
    Max. 7 %
  • Grain size < 100 mesh:
    Max. 1 %
  • CO₂-absorption capacity:
    Min. 19.0 %
  • Capacity:
    ≥19.0 % absorption capacity (CO₂)
  • Particle Size:
    +100 mesh

Specification Test Results

About this item

 :
Merck Index:13,08640
Storage Temperature:Ambient
UN:1907
ADR:8,III
 :GHS05
CAS Number:8006-28-8
MDL Number:MFCD00134094