Order Entry
Austria
ContactUsLinkComponent
 
n-Heptane 99%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
  650536-1L
 :  
undefined
n-Heptane 99%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
  650536-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Autoignition Temperature:
    433 °F
  • Purity (GC):
    Min. 98.5 %
  • IR spectrum:
    complying
  • Refractive index (n 20/D):
    1.3850 - 1.3890
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.01
  • Absorbance at 225 nm:
    Max. 0.10
  • Absorbance at 200 nm:
    Max. 1.0
  • Fluorescence (chinin) at 365 nm:
    Max. 1.0 ppb
  • GC (as heptachlor epoxide):
    Max. 2 µg/l
  • Vapor Density:
    3.5 (vs air)
  • Vapor Pressure:
    83 mmHg ( 37.7 °C)
  • Explosion Limit:
    7 %

 

 

 :
 :100.20 g/mol
 :13,04679
 :98.2…98.4 °C (1.000 hPa)
 :–90.5 °C
 :0.69 g/cm³ (15 °C)
 :6.7 %(V)
 :Ambient
 :205-563-8
 :1206
 :3,II
 :GHS07
 :GHS08
 :GHS02
 :GHS09
 :142-82-5
 :H₃C(CH₂)₅CH₃
 :MFCD00009544