Order Entry
Austria
ContactUsLinkComponent
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Corrosive
Irritant
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Identification:
    Passes test
  • Particle size (2 - 5 mm):
    Min. 78%
  • Absorption capacity (as CO2):
    Min. 19%
  • Loss on drying (105°C):
    13 - 18 %

Specification Test Results

Frequently Bought Together