ContactUsLinkComponent
Soda lime, pellets analytical reagent, with indicator
Corrosive
Irritant
Soda lime, pellets analytical reagent, with indicator
Supplier:  Merck
Certificates
Corrosive
Irritant
Soda lime, pellets analytical reagent, with indicator
Supplier:  Merck
Certificates
Specification Test Results
  • Identity:
    passes test
  • Absorption capacity for CO₂ (method NF XVII):
    ≥ 25 %
  • Loss on Drying (105°C):
    13.0 - 18.0 %
  • Color change:
    passes test
  • Bulk density:
    about 75
  • Particle size (2 - 5 mm):
    ≥ 95 %