Poly(caprolactone), M.W. ~10000 (GPC), flake or chunks, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 6-Caprolactone polymer, 2-Oxepanone homopolymer, PCL
CAS Number:24980-41-4
:
Formula:(C₆H₁₀O₂)n
MDL Number:MFCD00084404
Density:1.146 g/ml (25 °C)
Molecular Weight:114.1400 g/mol
Melting Pt:60 °C (lit.)
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Infrared spectrum:Conforms to structure
- Colour test (50 wt% xylene solution):≤150 APHA
- Viscosity (50 wt% xylene solution):400 - 1000 mPa.s
- Acid value (mg KOH/g):≤3.0
- Hydroxyl value (mg KOH/g):9.0 - 16.0
- Moecular weight (by GPC):Average Mn 10000
- Molecular weight:Average Mw 14000
Specification Test Results
Frequently Bought Together