Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Selenium, Sigma-Aldrich®

Toxic
Health
Supplier:   Merck
Selenium, Sigma-Aldrich®
Supplier:  Merck
Certificates
Selenium, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3283
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 59,20 €
    59,20 €
    1 * 5 g
       59,20 €

  • Glass bottle for solids
    20 g
    Log in for availability
    Log in for availability
    1 * 20 g
    1 * 20 g - 127,00 €
    127,00 €
    1 * 20 g
       127,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 306,00 €
    306,00 €
    1 * 100 g
       306,00 €