Tin ≥99.5% (metals basis), powder -100 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Boiling Pt:2270
Melting Pt:232
Density:7.31 g/cm³ (25 °C)
Molecular Weight:118.71 g/mol
Merck Index:12,09587
Storage Temperature:Ambient
EINECS:231-141-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together